accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 68298-46-4

H3CH3CNH2O

CAS RN 68298-46-4

IUPAC Name: 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-amine
Synonyms: 2,2-dimethyl-3H-1-benzofuran-7-amine
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: LMTIGABGABPAGU-UHFFFAOYSA-N
SMILES: CC1(C)CC2=CC=CC(N)=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results