accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NHNOH

CAS RN 68283-19-2

IUPAC Name: (2-butyl-1H-imidazol-5-yl)methanol
Synonyms: (2-butyl-3H-imidazol-4-yl)methanol
Molecular Weight (g/mol): 154.21
Molecular Formula: C8H14N2O
InChi Key: UZKBZGAMRJRWLR-UHFFFAOYSA-N
SMILES: CCCCC1=NC=C(CO)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results