accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOFFOOH3C

CAS RN 680-65-9

IUPAC Name: 1,3-diethyl 2,2-difluoropropanedioate
Synonyms: 1,3-diethyl 2,2-difluoropropanedioate
Molecular Weight (g/mol): 196.15
Molecular Formula: C7H10F2O4
InChi Key: WCRVWLHTROHXBB-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)C(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results