CAS RN 68-36-0
CAS RN 68-36-0
IUPAC Name:
1,4-bis(trichloromethyl)benzene
Synonyms:
P-DI(trichloromethyl)benzene
Molecular Weight (g/mol):
312.82
Molecular Formula:
C8H4Cl6
InChi Key:
OTEKOJQFKOIXMU-UHFFFAOYSA-N
SMILES:
ClC(Cl)(Cl)C1=CC=C(C=C1)C(Cl)(Cl)Cl
1
–
1
of
1
results
1
1,4-Bis(trichloromethyl)benzene, 99%, Thermo Scientific™
CAS: 68-36-0 Molecular Formula: C8H4Cl6 Molecular Weight (g/mol): 312.82 MDL Number: MFCD00000791 InChI Key: OTEKOJQFKOIXMU-UHFFFAOYSA-N Synonym: 1,4-bis trichloromethyl benzene,bitriben,chloksil,chloxil,chloxyl,hexichol,khloksil,khloxil,cloxil,hetol PubChem CID: 6233 IUPAC Name: 1,4-bis(trichloromethyl)benzene SMILES: ClC(Cl)(Cl)C1=CC=C(C=C1)C(Cl)(Cl)Cl