accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 68-34-8

CH3SOONH

CAS RN 68-34-8

IUPAC Name: 4-methyl-N-phenylbenzene-1-sulfonamide
Synonyms: p-toluenesulfonanilide
Molecular Weight (g/mol): 247.31
Molecular Formula: C13H13NO2S
InChi Key: VLVCWODDMDGANW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results