accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6797-44-0

IUPAC Name: (2E)-2-(N-hydroxyimino)-1-phenylbutane-1,3-dione
Synonyms: (2E)-2-(N-hydroxyimino)-1-phenylbutane-1,3-dione
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: JVPIIQSFMRUIAP-PKNBQFBNSA-N
SMILES: CC(=O)C(=N/O)\C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results