accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SSCH3H3CO

CAS RN 67952-60-7

IUPAC Name: 2-methyl-2-(methyldisulfanyl)propanal
Synonyms: 2-methyl-2-(methyldisulfanyl)propanal
Molecular Weight (g/mol): 150.25
Molecular Formula: C5H10OS2
InChi Key: VLBWEJJOETYCSE-UHFFFAOYSA-N
SMILES: CSSC(C)(C)C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results