accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6780-49-0

IUPAC Name: ethyl N-phenylcarboximidate
Synonyms: ethyl N-phenylcarboximidate
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: DRDBNKYFCOLNQO-UHFFFAOYSA-N
SMILES: CCOC=NC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results