accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 67627-18-3

IUPAC Name: tripotassium (Z)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5',7-trisulfonate
Synonyms: tripotassium (Z)-3,3'-dioxo-1H,1'H-[2,2'-biindolylidene]-5,5',7-trisulfonate
Molecular Weight (g/mol): 616.71
Molecular Formula: C16H7K3N2O11S3
InChi Key: XOSMXDUITYWYGR-JRYLAINFSA-K
SMILES: [K+].[K+].[K+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1/NC2=C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results