accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 67515-61-1

HOFFFF

CAS RN 67515-61-1

IUPAC Name: [4-fluoro-3-(trifluoromethyl)phenyl]methanol
Synonyms: [4-fluoro-3-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 194.13
Molecular Formula: C8H6F4O
InChi Key: DGBDVJVPBJMVDH-UHFFFAOYSA-N
SMILES: OCC1=CC=C(F)C(=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results