CAS RN 675126-10-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1S,4S)-N-Desmethyl Sertraline Hydrochloride, TRC
CAS: 675126-10-0 Molecular Formula: C16 H15 Cl2 N . Cl H Molecular Weight (g/mol): 328.66 Synonym: Norsertraline Hydrochloride,(1S,4S)-N-Desmethylsertraline Hydrochloride IUPAC Name: (1S,4S)-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride SMILES: Cl.N[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13