accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6750-04-5

OOCH

CAS RN 6750-04-5

IUPAC Name: prop-2-yn-1-yl benzoate
Synonyms: prop-2-yn-1-yl benzoate
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: NBDHEMWCIUHARG-UHFFFAOYSA-N
SMILES: O=C(OCC#C)C1=CC=CC=C1
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)