accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 67442-07-3

H3CONCH3OCl

CAS RN 67442-07-3

IUPAC Name: 2-chloro-N-methoxy-N-methylacetamide
Synonyms: 2-chloro-N-methoxy-N-methylacetamide
Molecular Weight (g/mol): 137.56
Molecular Formula: C4H8ClNO2
InChi Key: SCOJKGRNQDKFRP-UHFFFAOYSA-N
SMILES: CON(C)C(=O)CCl
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)