accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6728-26-3

CH3O(E)

CAS RN 6728-26-3

IUPAC Name: (2E)-hex-2-enal
Synonyms: (E)-2-hexenal 3-propyl acrolein 2-hexenal(trans)
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: MBDOYVRWFFCFHM-SNAWJCMRSA-N
SMILES: CCC\C=C\C=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)