accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 672325-37-0

IUPAC Name: 1-[3-(4-methylpiperazin-1-yl)phenyl]methanamine
Synonyms: 1-[3-(4-methylpiperazin-1-yl)phenyl]methanamine
Molecular Weight (g/mol): 205.31
Molecular Formula: C12H19N3
InChi Key: VKTLTRIKVYJVOX-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC=CC(CN)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results