accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 67169-22-6

CH3FFFNH2

CAS RN 67169-22-6

IUPAC Name: 2-methyl-4-(trifluoromethyl)aniline
Synonyms: 2-methyl-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 175.15
Molecular Formula: C8H8F3N
InChi Key: PAXQXJDYVORMOO-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC(=C1)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results