accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 66946-48-3

SSSSSSSS

CAS RN 66946-48-3

IUPAC Name: 2-{2H,5H,6H-[1,3]dithiolo[4,5-b][1,4]dithiin-2-ylidene}-2H,5H,6H-[1,3]dithiolo[4,5-b][1,4]dithiine
Synonyms: 2-{5H,6H-[1,3]dithiolo[4,5-b][1,4]dithiin-2-ylidene}-5H,6H-[1,3]dithiolo[4,5-b][1,4]dithiine
Molecular Weight (g/mol): 384.65
Molecular Formula: C10H8S8
InChi Key: LZJCVNLYDXCIBG-UHFFFAOYSA-N
SMILES: C1CSC2=C(SC(S2)=C2SC3=C(S2)SCCS3)S1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results