CAS RN 66937-99-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(±)-Morpholine-3-carboxylic acid hydrochloride, 96%, Thermo Scientific™
CAS: 66937-99-3 Molecular Formula: C5H9NO3 Molecular Weight (g/mol): 131.13 MDL Number: MFCD01318649,MFCD03426306,MFCD03426307 InChI Key: JUNOWSHJELIDQP-UHFFFAOYNA-N Synonym: 3-morpholinecarboxylic acid,3-morpholine carboxylic acid,r-3-morpholinecarboxylic acid,3-morpholinecarboxylicacid,3-morpholinecarboxylicacid, 3r,3-morpholinecarboxylicacid, 3s,acmc-20agwb,acmc-209pcc,acmc-209vok,3-morphline carboxylic acid PubChem CID: 3340152 IUPAC Name: morpholine-3-carboxylic acid SMILES: OC(=O)C1COCCN1