accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3OONNNSOHO

CAS RN 668484-45-5

IUPAC Name: 2-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}-1,3-thiazole-4-carboxylic acid
Synonyms: 2-[4-(tert-butoxycarbonyl)piperazin-1-yl]-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 313.37
Molecular Formula: C13H19N3O4S
InChi Key: FMSFMUBALWPCQM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=NC(=CS1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results