CAS RN 668273-75-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Boc Linagliptin, TRC
CAS: 668273-75-4 Molecular Formula: C30 H36 N8 O4 Molecular Weight (g/mol): 572.66 Synonym: N-[(3R)-1-[7-(2-Butyn-1-yl)-2,3,6,7-tetrahydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-2,6-dioxo-1H-purin-8-yl]-3-piperidinyl]carbamic Acid 1,1-Dimethylethyl Ester IUPAC Name: tert-butyl N-[(3R)-1-[7-but-2-ynyl-3-methyl-1-[(4-methylquinazolin-2-yl)methyl]-2,6-dioxopurin-8-yl]piperidin-3-yl]carbamate SMILES: CC#CCn1c(nc2N(C)C(=O)N(Cc3nc(C)c4ccccc4n3)C(=O)c12)N5CCC[C@H](C5)NC(=O)OC(C)(C)C