accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 66674-16-6

CH3(S)OHNHOO

CAS RN 66674-16-6

IUPAC Name: benzyl N-(1-hydroxypropan-2-yl)carbamate
Synonyms: benzyl N-(1-hydroxypropan-2-yl)carbamate
Molecular Weight (g/mol): 209.25
Molecular Formula: C11H15NO3
InChi Key: AFPHMHSLDRPUSM-UHFFFAOYNA-N
SMILES: CC(CO)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)