CAS RN 66505-80-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Nitrosodiclofenac, TRC
CAS: 66505-80-4 Molecular Formula: C14H10Cl2N2O3 Molecular Weight (g/mol): 325.15 Synonym: Benzeneacetic acid, 2-[(2,6-dichlorophenyl)nitrosoamino]-,2-[(2,6-Dichlorophenyl)nitrosoamino]benzeneacetic acid,N-Nitrosodiclofenac,NCX 284,2-[(2,6-Dichlorophenyl)nitrosoamino]benzeneacetic Acid,NCX 284 IUPAC Name: 2-[2-(2,6-dichloro-N-nitrosoanilino)phenyl]acetic acid SMILES: OC(=O)Cc1ccccc1N(N=O)c2c(Cl)cccc2Cl