CAS RN 66487-95-4
CAS RN 66487-95-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dimethyl Fumarate-d6, TRC
CAS: 66487-95-4 Molecular Formula: C6H2D6O4 Molecular Weight (g/mol): 150.16 Synonym: (2E)-2-Butenedioic Acid 1,4-Dimethyl Ester-d6,(E)-2-Butenedioic Acid 1,4-Dimethyl Ester-d6,trans-1,2-Ethylenedicarboxylic Acid,Fumaric Acid Dimethyl Ester-d6,Di(methyl-d3) Ester (2E)-2-Butenedioic Acid,Di(methyl-d3) Ester (E)-2-Butenedioic Acid SMILES: [2H]C([2H])([2H])OC(=O)\C=C\C(=O)OC([2H])([2H])[2H]