accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 661465-44-7

BClFFFK

CAS RN 661465-44-7

IUPAC Name: potassium (4-chlorophenyl)trifluoroboranuide
Synonyms: potassium (4-chlorophenyl)trifluoroboranuide
Molecular Weight (g/mol): 218.45
Molecular Formula: C6H4BClF3K
InChi Key: GXBSKMLVYJPMRT-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results