accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OO

CAS RN 66109-88-4

IUPAC Name: 4-phenyloxane-4-carbaldehyde
Synonyms: 4-phenyloxane-4-carbaldehyde
Molecular Weight (g/mol): 190.24
Molecular Formula: C12H14O2
InChi Key: IEIVCCVSMYLGOC-UHFFFAOYSA-N
SMILES: O=CC1(CCOCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results