accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6590-94-9

ClClNCS

CAS RN 6590-94-9

IUPAC Name: 1,2-dichloro-4-isothiocyanatobenzene
Synonyms: 1,2-dichloro-4-isothiocyanatobenzene
Molecular Weight (g/mol): 204.07
Molecular Formula: C7H3Cl2NS
InChi Key: OSBIEFWIIINTNJ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1Cl)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results