accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2ONHF

CAS RN 659-30-3

IUPAC Name: (4-fluorophenyl)urea
Synonyms: P-fluorophenylurea
Molecular Weight (g/mol): 154.14
Molecular Formula: C7H7FN2O
InChi Key: IQZBVVPYTDHTIP-UHFFFAOYSA-N
SMILES: NC(=O)NC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results