CAS RN 6581-66-4
IUPAC Name:
2-methoxyoxane
Synonyms:
pyran, 2-methoxy-tetrahydro-
Molecular Weight (g/mol):
116.16
Molecular Formula:
C6H12O2
InChi Key:
XTDKZSUYCXHXJM-UHFFFAOYNA-N
SMILES:
COC1CCCCO1
1
–
1
of
1
results
1
2-Methoxytetrahydropyran, 98%
CAS: 6581-66-4 Molecular Formula: C6H12O2 Molecular Weight (g/mol): 116.16 InChI Key: XTDKZSUYCXHXJM-UHFFFAOYSA-N Synonym: 2-methoxytetrahydropyran,tetrahydro-2-methoxy-2h-pyran,pyran, 2-methoxy-tetrahydro,2h-pyran,tetrahydro-2-methoxy,2h-pyran, tetrahydro-2-methoxy,2-methoxytetrahydro-2h-pyran,methoxytetrahydropyran,2-methoxyoxan-3-yl,5-17-03-00078 beilstein handbook reference PubChem CID: 23057 IUPAC Name: 2-methoxyoxane SMILES: COC1CCCCO1