accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2ClN

CAS RN 6575-11-7

IUPAC Name: 2-amino-6-chlorobenzonitrile
Synonyms: 2-amino-6-chlorobenzonitrile
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: MEJVTQKBWPYBFG-UHFFFAOYSA-N
SMILES: NC1=CC=CC(Cl)=C1C#N
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)