accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFClClO

CAS RN 657-05-6

IUPAC Name: 2-chloro-5-(trifluoromethyl)benzoyl chloride
Synonyms: 2-chloro-5-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 243.01
Molecular Formula: C8H3Cl2F3O
InChi Key: FRARPACTAFPNNL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Cl)C(=C1)C(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results