accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 65515-28-8

CH3OONClCl

CAS RN 65515-28-8

IUPAC Name: methyl 2,6-dichloropyridine-3-carboxylate
Synonyms: methyl 2,6-dichloropyridine-3-carboxylate
Molecular Weight (g/mol): 206.02
Molecular Formula: C7H5Cl2NO2
InChi Key: IFVVGOJYWCHRCT-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)N=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results