accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOFFFFF

CAS RN 653-21-4

IUPAC Name: 2-(2,3,4,5,6-pentafluorophenyl)acetic acid
Synonyms: pentafluorophenylacetic acid
Molecular Weight (g/mol): 226.1
Molecular Formula: C8H3F5O2
InChi Key: LGCODSNZJOVMHV-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results