CAS RN 6526-67-6
IUPAC Name:
2-chloro-N,N-dimethylbenzamide
Synonyms:
2-chloro-N,N-dimethylbenzamide
Molecular Weight (g/mol):
183.64
Molecular Formula:
C9H10ClNO
InChi Key:
SZUQPOLAEFBJJH-UHFFFAOYSA-N
SMILES:
CN(C)C(=O)C1=CC=CC=C1Cl
1
–
1
of
1
results
1
2-Chloro-N,N-dimethylbenzamide, 97%, Thermo Scientific Chemicals
CAS: 6526-67-6 Molecular Formula: C9H10ClNO Molecular Weight (g/mol): 183.64 MDL Number: MFCD00463636 InChI Key: SZUQPOLAEFBJJH-UHFFFAOYSA-N Synonym: benzamide, 2-chloro-n,n-dimethyl,n,n-dimethyl-2-chlorobenzamide,o-chloro-n,n-dimethylbenzamide,2-chloro-n,n-dimethyl-benzamide,benzamide, o-chloro-n,n-dimethyl,aronis25704,2-chlorophenyl-n,n-dimethylcarboxamide PubChem CID: 81012 IUPAC Name: 2-chloro-N,N-dimethylbenzamide SMILES: CN(C)C(=O)C1=CC=CC=C1Cl