CAS RN 65208-41-5
IUPAC Name:
2-sulfanylacetic acid calcium trihydrate
Synonyms:
thioglycolic acid calcium trihydrate
Molecular Weight (g/mol):
186.24
Molecular Formula:
C2H10CaO5S
InChi Key:
GDECCIJZCWLFSB-UHFFFAOYSA-N
SMILES:
O.O.O.[Ca].OC(=O)CS
1
–
3
of
3
results
1
Calcium thioglycolate trihydrate, 99%
CAS: 65208-41-5 Molecular Formula: C2H10CaO5S Molecular Weight (g/mol): 186.24 MDL Number: MFCD00150707 InChI Key: GDECCIJZCWLFSB-UHFFFAOYSA-N Synonym: calciumthioglycolatetrihydrate PubChem CID: 131881455 SMILES: O.O.O.[Ca].OC(=O)CS
2
Mercaptoacetic acid calcium salt trihydrate, 98%, Thermo Scientific™
CAS: 65208-41-5 Molecular Formula: C2H10CaO5S Molecular Weight (g/mol): 186.24 MDL Number: MFCD00150707 InChI Key: GDECCIJZCWLFSB-UHFFFAOYSA-N Synonym: calcium,2-sulfidoacetate,trihydrate,thioglycolic acid calcium trihydrate PubChem CID: 131882808 IUPAC Name: calcium;2-sulfanylacetic acid;trihydrate SMILES: O.O.O.[Ca].OC(=O)CS