accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NaNaCH3SOOONNCH3NHNOSOOOSOOOCH3(E/Z)(E)

CAS RN 6459-94-5

IUPAC Name: disodium (8E)-8-{2-[3,3'-dimethyl-4'-(2-{4-[(4-methylbenzenesulfonyl)oxy]phenyl}diazen-1-yl)-[1,1'-biphenyl]-4-yl]hydrazin-1-ylidene}-7-oxo-7,8-dihydronaphthalene-1,3-disulfonate
Synonyms: disodium (8E)-8-{2-[3,3'-dimethyl-4'-(2-{4-[(4-methylbenzenesulfonyl)oxy]phenyl}diazen-1-yl)-[1,1'-biphenyl]-4-yl]hydrazin-1-ylidene}-7-oxonaphthalene-1,3-disulfonate
Molecular Weight (g/mol): 830.81
Molecular Formula: C37H28N4Na2O10S3
InChi Key: PVLKQJLDEXCKNY-JYLPYTDFSA-L
SMILES: [Na+].[Na+].CC1=CC=C(C=C1)S(=O)(=O)OC1=CC=C(C=C1)N=NC1=CC=C(C=C1C)C1=CC=C(N\N=C2\C(=O)C=CC3=CC(=CC(=C23)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results