accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6456-74-2

IUPAC Name: tert-butyl 2-aminoacetate
Synonyms: tert-butyl 2-aminoacetate
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: SJMDMGHPMLKLHQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results