accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 64438-54-6

GdOOOOOOCCCH3CH3CCH3H3CCH3H3CHHH

CAS RN 64438-54-6

IUPAC Name: gadolinium(3+) tris(4-oxopent-2-en-2-olate) hydrate
Synonyms: gadolinium(3+) tris(4-oxopent-2-en-2-olate) hydrate
Molecular Weight (g/mol): 472.59
Molecular Formula: C15H23GdO7
InChi Key: NJOCOKMBKFILAF-UHFFFAOYSA-K
SMILES: O.[Gd+3].CC([O-])=CC(C)=O.CC([O-])=CC(C)=O.CC([O-])=CC(C)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results