accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 64438-53-5

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHSm

CAS RN 64438-53-5

IUPAC Name: tris(4-hydroxypent-3-en-2-one) hydrate samarium
Synonyms: tris(4-hydroxypent-3-en-2-one) hydrate samarium
Molecular Weight (g/mol): 468.73
Molecular Formula: C15H26O7Sm
InChi Key: FULVMVWDCYMZRZ-UHFFFAOYSA-N
SMILES: O.[Sm].CC(O)=CC(C)=O.CC(O)=CC(C)=O.CC(O)=CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results