accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 64424-12-0

LaOOOOOOCCCH3CH3CCH3H3CCH3H3CHHH

CAS RN 64424-12-0

IUPAC Name: tris(4-hydroxypent-3-en-2-one) dihydrate lanthanum
Synonyms: tris(4-hydroxypent-3-en-2-one) dihydrate lanthanum
Molecular Weight (g/mol): 475.29
Molecular Formula: C15H28LaO8
InChi Key: DTGMYUJFAPMLCR-UHFFFAOYSA-N
SMILES: O.O.[La].CC(O)=CC(C)=O.CC(O)=CC(C)=O.CC(O)=CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results