accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOSNNBr

CAS RN 6439-91-4

IUPAC Name: ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate
Synonyms: ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate
Molecular Weight (g/mol): 237.07
Molecular Formula: C5H5BrN2O2S
InChi Key: MBEUYMRLYKXBGT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(Br)SN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results