accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CBrNH2HCl

CAS RN 64265-77-6

IUPAC Name: 1-(4-bromophenyl)ethan-1-amine hydrochloride
Synonyms: 1-(4-bromophenyl)ethanamine hydrochloride
Molecular Weight (g/mol): 236.54
Molecular Formula: C8H11BrClN
InChi Key: BQCAANUXMMQVAY-UHFFFAOYNA-N
SMILES: Cl.CC(N)C1=CC=C(Br)C=C1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results