CAS RN 64262-23-3
CAS RN 64262-23-3
IUPAC Name:
dihydrogen 1-propylpiperazine dibromide
Synonyms:
dihydrogen 1-propylpiperazine dibromide
Molecular Weight (g/mol):
290.04
Molecular Formula:
C7H18Br2N2
InChi Key:
YTPQLWVHCBATKO-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Br-].[Br-].CCCN1CCNCC1
1
–
1
of
1
results
1
1-n-Propylpiperazine dihydrobromide, 98%, Thermo Scientific™
CAS: 64262-23-3 Molecular Formula: C7H18Br2N2 Molecular Weight (g/mol): 290.043 MDL Number: MFCD00051992 InChI Key: YTPQLWVHCBATKO-UHFFFAOYSA-N Synonym: 1-propylpiperazine dihydrobromide,1-n-propylpiperazine dihydrobromide,n-propylpiperazinium dibromide,n-propylpiperazine dihydrobromide,1-n-propylpiperazine 2hbr,piperazine, 1-propyl-, dihydrobromide,acmc-209nl5,ksc494c5l,propylpiperazine, bromide, bromide,1-n-propyl-piperazine dihydrobromide PubChem CID: 3085288 IUPAC Name: 1-propylpiperazine;dihydrobromide SMILES: CCCN1CCNCC1.Br.Br