accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2N(S)NHOO

CAS RN 64090-98-8

IUPAC Name: (2S)-2-amino-3-(pyridin-3-yl)propanoic acid
Synonyms: (2S)-2-amino-3-(pyridin-3-yl)propanoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: DFZVZEMNPGABKO-ZETCQYMHSA-N
SMILES: N[C@@H](CC1=CC=CN=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results