accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 64030-44-0

NHHN

CAS RN 64030-44-0

IUPAC Name: N-[(pyrrolidin-2-yl)methyl]aniline
Synonyms: N-(pyrrolidin-2-ylmethyl)aniline
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: MCHWKJRTMPIHRA-UHFFFAOYNA-N
SMILES: C(NC1=CC=CC=C1)C1CCCN1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results