accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClH3NOO

CAS RN 63989-79-7

IUPAC Name: 2-oxo-2H-chromen-6-aminium chloride
Synonyms: 2-oxochromen-6-aminium chloride
Molecular Weight (g/mol): 197.62
Molecular Formula: C9H8ClNO2
InChi Key: OSIGAIXSSYAHEG-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1=CC=C2OC(=O)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results