accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 63843-46-9

CH3ONH

CAS RN 63843-46-9

IUPAC Name: 4-(3-methylphenoxy)piperidine
Synonyms: 4-(3-methylphenoxy)piperidine
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: RTSXWJCYCFVWBU-UHFFFAOYSA-N
SMILES: CC1=CC(OC2CCNCC2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results