CAS RN 63799-11-1
IUPAC Name:
4'-[(2S)-2-methylbutyl]-[1,1'-biphenyl]-4-carbonitrile
Synonyms:
4'-[(2S)-2-methylbutyl]-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol):
249.36
Molecular Formula:
C18H19N
InChi Key:
DNJQGRFZQMOYGM-AWEZNQCLSA-N
SMILES:
CC[C@H](C)CC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl, 97%
CAS: 63799-11-1 Molecular Formula: C18H19N Molecular Weight (g/mol): 249.357 MDL Number: MFCD00192280 InChI Key: DNJQGRFZQMOYGM-AWEZNQCLSA-N Synonym: s-4'-2-methylbutyl-1,1'-biphenyl-4-carbonitrile,s-4-cyano-4'-2-methylbutyl biphenyl,4-s-2-methylbutyl-4'-cyanobiphenyl,s-4'-2-methylbutyl 1,1'-biphenyl-4-carbonitrile,4-4-2s-2-methylbutyl phenyl benzonitrile,1,1'-biphenyl-4-carbonitrile,4'-2s-2-methylbutyl,1,1'-biphenyl-4-carbonitrile, 4'-2s-2-methylbutyl,4-cyano-4'-2-methylbut-1-yl biphenyl PubChem CID: 114240 IUPAC Name: 4-[4-[(2S)-2-methylbutyl]phenyl]benzonitrile SMILES: CCC(C)CC1=CC=C(C=C1)C2=CC=C(C=C2)C#N