accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6373-50-8

CH3SNHONHClClNN

CAS RN 6373-50-8

IUPAC Name: 1-(2,4-dichlorophenyl)-3-[5-(4-methylphenyl)-1,3,4-thiadiazol-2-yl]urea
Synonyms: 1-(2,4-dichlorophenyl)-3-[5-(4-methylphenyl)-1,3,4-thiadiazol-2-yl]urea
Molecular Weight (g/mol): 379.26
Molecular Formula: C16H12Cl2N4OS
InChi Key: YDTTUXLKBBQRHA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=NN=C(NC(=O)NC2=CC=C(Cl)C=C2Cl)S1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results