accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HClCH3HNNH2

CAS RN 637-04-7

IUPAC Name: hydrogen (3-methylphenyl)hydrazine chloride
Synonyms: hydrogen (3-methylphenyl)hydrazine chloride
Molecular Weight (g/mol): 158.63
Molecular Formula: C7H11ClN2
InChi Key: SHTBTGXNVWYODU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=CC(NN)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)