accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 637-03-6

IUPAC Name: arsorosobenzene
Synonyms: oxophenylarsine
Molecular Weight (g/mol): 168.03
Molecular Formula: C6H5AsO
InChi Key: BQVCCPGCDUSGOE-UHFFFAOYSA-N
SMILES: O=[As]C1=CC=CC=C1
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results